accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OFFFOHO

CAS RN 213598-09-5

IUPAC Name: 4-methoxy-3-(trifluoromethyl)benzoic acid
Synonyms: 4-methoxy-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: VNWUPESTVBTYJM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results