accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21331-43-1

IUPAC Name: 4-(naphthalen-2-yl)-1,3-thiazol-2-amine
Synonyms: 4-(naphthalen-2-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 226.3
Molecular Formula: C13H10N2S
InChi Key: GWDNDNTTXIIXRS-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results