accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2122-63-6

NH2IOHO

CAS RN 2122-63-6

IUPAC Name: 4-amino-3-iodobenzoic acid
Synonyms: benzoic acid, 4-amino-3-iodo-
Molecular Weight (g/mol): 263.03
Molecular Formula: C7H6INO2
InChi Key: FPLDDZRJTOXFCB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1I)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results