CAS RN 21109-95-5
IUPAC Name:
barium(2+) sulfanediide
Synonyms:
BaS barium sulfide bariumsulfid barium sulphide
Molecular Weight (g/mol):
169.39
Molecular Formula:
BaS
InChi Key:
CJDPJFRMHVXWPT-UHFFFAOYSA-N
SMILES:
[S--].[Ba++]
1
–
1
of
1
results
1
Barium sulfide, 99.7% (metals basis)
CAS: 21109-95-5 Molecular Formula: BaS Molecular Weight (g/mol): 169.39 MDL Number: MFCD00014192 InChI Key: CJDPJFRMHVXWPT-UHFFFAOYSA-N Synonym: barium sulfide,thioxobarium PubChem CID: 88791 IUPAC Name: barium(2+) sulfanediide SMILES: [S--].[Ba++]