CAS RN 210421-74-2
CAS RN 210421-74-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sitaxsentan sodium, MedChemExpress
MedChemExpress Sitaxsentan sodium (IPI 1040 sodium; TBC11251 sodium) is an orally active, highly selective antagonist of endothelin A receptors.