accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2103-88-0

SNHS

CAS RN 2103-88-0

IUPAC Name: 4-phenyl-2,3-dihydro-1,3-thiazole-2-thione
Synonyms: 4-phenyl-3H-1,3-thiazole-2-thione
Molecular Weight (g/mol): 193.28
Molecular Formula: C9H7NS2
InChi Key: CYCKHTAVNBPQDB-UHFFFAOYSA-N
SMILES: S=C1NC(=CS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results