accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOH

CAS RN 21021-55-6

IUPAC Name: 4-dodecylbenzoic acid
Synonyms: 4-dodecylbenzoic acid
Molecular Weight (g/mol): 290.45
Molecular Formula: C19H30O2
InChi Key: JCHCIOJFCCQEKZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results