accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2100-17-6

IUPAC Name: pent-4-enal
Synonyms: 4-pentenal
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: QUMSUJWRUHPEEJ-UHFFFAOYSA-N
SMILES: C=CCCC=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results