CAS RN 2098836-54-3
CAS RN 2098836-54-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ VH 032, thiol
Functionalized VHL ligand for PROTAC™; development
2
VH032 thiol, MedChemExpress
MedChemExpress VH032 thiol (VHL ligand 6) is a VHL ligand, which binds to pan-BET inhibitor JQ1 via a linker to form PROTAC.