accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2096341-27-2

BOHNNOOClHOOH

CAS RN 2096341-27-2

IUPAC Name: {2-chloro-5-[(3-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-5-[(3-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 320.49
Molecular Formula: C13H10BClN2O5
InChi Key: LEIFVWJAWHHHHN-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(=O)NC1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results