accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONNOOHOO

CAS RN 209593-18-0

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperazine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-[(9H-fluoren-9-ylmethoxy)carbonyl]piperazine-2-carboxylic acid
Molecular Weight (g/mol): 452.51
Molecular Formula: C25H28N2O6
InChi Key: ZVHNNCSUTNWKFC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results