accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2088-24-6

IUPAC Name: 2-(3-methoxyphenoxy)acetic acid
Synonyms: M-methoxyphenoxyacetic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: AHDPQRIYMMZJTF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OCC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results