accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20876-37-3

CH3ONOO

CAS RN 20876-37-3

IUPAC Name: 1-(benzyloxy)-2-methyl-3-nitrobenzene
Synonyms: 1-(benzyloxy)-2-methyl-3-nitrobenzene
Molecular Weight (g/mol): 243.26
Molecular Formula: C14H13NO3
InChi Key: PBSZHNXXFIYDBU-UHFFFAOYSA-N
SMILES: CC1=C(OCC2=CC=CC=C2)C=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results