accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20857-47-0

IUPAC Name: 3,5-bis(trifluoromethyl)pyridine
Synonyms: 3,5-bis(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.1
Molecular Formula: C7H3F6N
InChi Key: RQEOYYWUVYZZLL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CN=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results