accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20824-56-0

NH4NH4HOONNOHOOOOO

CAS RN 20824-56-0

IUPAC Name: diammonium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Synonyms: diammonium edta
Molecular Weight (g/mol): 326.31
Molecular Formula: C10H22N4O8
InChi Key: KYQODXQIAJFKPH-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results