accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20806-43-3

IUPAC Name: 3-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Synonyms: 3-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 329.35
Molecular Formula: C18H19NO5
InChi Key: CYYRLHUAMWRBHC-UHFFFAOYNA-N
SMILES: OC(=O)C(COCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results