accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 207981-45-1

OHFFFF

CAS RN 207981-45-1

IUPAC Name: [2-fluoro-3-(trifluoromethyl)phenyl]methanol
Synonyms: [2-fluoro-3-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: KKHMGVIWWRPCNH-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results