CAS RN 207861-63-0
IUPAC Name:
nickel(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms:
nickel(2+) bis(bistriflylimide anion)
Molecular Weight (g/mol):
618.96
Molecular Formula:
C4F12N2NiO8S4
InChi Key:
VFUSIQAHVPEYFD-UHFFFAOYSA-N
SMILES:
[Ni++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Nickel bis(trifluoromethylsulfonyl)imide
CAS: 207861-63-0 Molecular Formula: C4F12N2NiO8S4 Molecular Weight (g/mol): 618.964 MDL Number: MFCD23380174 InChI Key: VFUSIQAHVPEYFD-UHFFFAOYSA-N Synonym: nickel bis-trifluoromethanesulfonimide PubChem CID: 131875825 IUPAC Name: bis(trifluoromethylsulfonyl)azanide;nickel(2+) SMILES: C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]