accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrOOHH2N

CAS RN 20776-48-1

IUPAC Name: 2-amino-6-bromobenzoic acid
Synonyms: 2-amino-6-bromobenzoic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: BNQPROAXWQCNKO-UHFFFAOYSA-N
SMILES: NC1=C(C(O)=O)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results