CAS RN 207683-21-4
CAS RN 207683-21-4
IUPAC Name:
disodium tetrachloroplatinumdiuide hydrate
Synonyms:
disodium tetrachloroplatinate(2-) hydrate
Molecular Weight (g/mol):
400.88
Molecular Formula:
Cl4H2Na2OPt
InChi Key:
GIEPPPFPHULPBK-UHFFFAOYSA-J
SMILES:
O.[Na+].[Na+].Cl[Pt--](Cl)(Cl)Cl
1
–
1
of
1
results
1
Sodium tetrachloroplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 42.4% min
CAS: 207683-21-4 Molecular Formula: Cl4Na2Pt Molecular Weight (g/mol): 382.87 MDL Number: MFCD00150188 InChI Key: DOAIHRYZKUCMDX-UHFFFAOYSA-J Synonym: sodium tetrachloroplatinate ii hydrate,disodium tetrachloroplatinate 2-hydrate,sodium tetrachloroplatinate 2--water 2/1/1 PubChem CID: 56846332 IUPAC Name: disodium;tetrachloroplatinum(2-);hydrate SMILES: [Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--]