accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 207572-67-6

CH3NNHNSOOHOSOOHOOOOPOHOOHNaNaNaNa(E/Z)

CAS RN 207572-67-6

IUPAC Name: 2-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonooxy)methyl]-2,5-dihydropyridin-2-ylidene}hydrazin-1-yl)benzene-1,4-disulfonic acid tetrasodium
Synonyms: 2-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonooxy)methyl]pyridin-2-ylidene}hydrazin-1-yl)benzene-1,4-disulfonic acid tetrasodium
Molecular Weight (g/mol): 603.33
Molecular Formula: C14H14N3Na4O12PS2
InChi Key: XPRWSDRXOGVWIR-UHFFFAOYSA-N
SMILES: [Na].[Na].[Na].[Na].CC1=NC(=NNC2=C(C=CC(=C2)S(O)(=O)=O)S(O)(=O)=O)C(COP(O)(O)=O)=C(C=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results