accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrBrNi

CAS RN 207569-11-7

IUPAC Name: dibromonickel hydrate
Synonyms: dibromonickel hydrate
Molecular Weight (g/mol): 236.52
Molecular Formula: Br2H2NiO
InChi Key: LQJMXNQEJAVYNB-UHFFFAOYSA-L
SMILES: O.Br[Ni]Br
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)