accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20739-58-6

H3COH

CAS RN 20739-58-6

IUPAC Name: oct-2-yn-1-ol
Synonyms: 2-octyn-1-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: TTWYFVOMGMBZCF-UHFFFAOYSA-N
SMILES: CCCCCC#CCO
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)