accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SOOOH2ONa

CAS RN 207300-91-2

IUPAC Name: sodium hexane-1-sulfonate hydrate
Synonyms: sodium hexane-1-sulfonate hydrate
Molecular Weight (g/mol): 206.23
Molecular Formula: C6H15NaO4S
InChi Key: CLCGFJYKZGFGSQ-UHFFFAOYSA-M
SMILES: O.[Na+].CCCCCCS([O-])(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results