CAS RN 207300-70-7
CAS RN 207300-70-7
IUPAC Name:
sodium 2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
Synonyms:
sodium 2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
Molecular Weight (g/mol):
234.14
Molecular Formula:
C6H11NaO8
InChi Key:
ICQRTPLATMRWJI-UHFFFAOYNA-M
SMILES:
O.[Na+].OC(C=O)C(O)C(O)C(O)C([O-])=O
1
–
1
of
1
results
1
Promotions Available
Sodium D-glucuronate monohydrate, 99%
Sodium D-glucuronate monohydrate, 99%
CAS: 207300-70-7 Molecular Formula: C6H11NaO8 Molecular Weight (g/mol): 234.14 MDL Number: MFCD00151051 InChI Key: ICQRTPLATMRWJI-UHFFFAOYNA-M Synonym: d-glucuronic acid sodium salt hydrate,glv8d2bupd,unii-glv8d2bupd,sodium d-glucuronate,sodium d-glucuronate monohydrate,sodium glucuronate hydrate jan,sodium 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate,sodium-d-glucuronate,d-glucuronic acid sodium salt PubChem CID: 24892836 SMILES: O.[Na+].OC(C=O)C(O)C(O)C(O)C([O-])=O