accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OSOO(R)HSHS

CAS RN 207233-91-8

IUPAC Name: (2R)-2,3-disulfanylpropane-1-sulfonate
Synonyms: (2R)-2,3-disulfanylpropane-1-sulfonate
Molecular Weight (g/mol): 187.27
Molecular Formula: C3H7O3S3
InChi Key: JLVSRWOIZZXQAD-GSVOUGTGSA-M
SMILES: [O-]S(=O)(=O)C[C@H](S)CS
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)