accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 206996-61-4

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHCe

CAS RN 206996-61-4

IUPAC Name: cerium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Synonyms: cerium(3+) tris(4-oxopent-2-en-2-olate) hydrate
Molecular Weight (g/mol): 455.46
Molecular Formula: C15H23CeO7
InChi Key: KGJWXERBZWXYFB-UHFFFAOYSA-K
SMILES: O.[Ce+3].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results