accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 206551-41-9

CH3OOBrF

CAS RN 206551-41-9

IUPAC Name: methyl 3-bromo-2-fluorobenzoate
Synonyms: methyl 3-bromo-2-fluorobenzoate
Molecular Weight (g/mol): 233.04
Molecular Formula: C8H6BrFO2
InChi Key: ZWOFHFOFKBYRHV-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results