accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20603-00-3

IUPAC Name: 2-(azepan-1-yl)ethan-1-ol
Synonyms: 2-(azepan-1-yl)ethanol
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: VMRYMOMQCYSPHS-UHFFFAOYSA-N
SMILES: OCCN1CCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results