accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20583-66-8

OFFFFFFFOFFF

CAS RN 20583-66-8

IUPAC Name: 1,1,1,5,5,6,6,7,7,7-decafluoroheptane-2,4-dione
Synonyms: 1,1,1,5,5,6,6,7,7,7-decafluoroheptane-2,4-dione
Molecular Weight (g/mol): 308.08
Molecular Formula: C7H2F10O2
InChi Key: SUORUQZBFOQDGX-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC(=O)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results