accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20583-31-7

IUPAC Name: 5-(4-nitrophenyl)-1H-pyrazole
Synonyms: 3-(4-nitrophenyl)-2H-pyrazole
Molecular Weight (g/mol): 189.17
Molecular Formula: C9H7N3O2
InChi Key: IPIYADCDDIUVPS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results