accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2055752-24-2

BOOH3CCH3H3CH3CNN

CAS RN 2055752-24-2

IUPAC Name: 1-phenyl-4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine
Synonyms: 1-phenyl-4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine
Molecular Weight (g/mol): 378.32
Molecular Formula: C23H31BN2O2
InChi Key: YIEPQKWTXHUCQJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCN(CC2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results