accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SOONH2

CAS RN 20503-40-6

IUPAC Name: 6-amino-1λ⁶-benzothiophene-1,1-dione
Synonyms: 6-amino-1λ⁶-benzothiophene-1,1-dione
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: KRUCRVZSHWOMHC-UHFFFAOYSA-N
SMILES: NC1=CC2=C(C=CS2(=O)=O)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)