CAS RN 2049-96-9
IUPAC Name:
pentyl benzoate
Synonyms:
amyl benzoate
Molecular Weight (g/mol):
192.26
Molecular Formula:
C12H16O2
InChi Key:
QKNZNUNCDJZTCH-UHFFFAOYSA-N
SMILES:
CCCCCOC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
n-Pentyl benzoate, 98+%
CAS: 2049-96-9 Molecular Formula: C12H16O2 Molecular Weight (g/mol): 192.258 MDL Number: MFCD00048843 InChI Key: QKNZNUNCDJZTCH-UHFFFAOYSA-N Synonym: amyl benzoate,n-pentyl benzoate,benzoic acid, pentyl ester,n-amyl benzoate,unii-cle7k4b29s,cle7k4b29s,amylbenzoat,amylbenzoate,n amyl benxoate,benzoic acid pentyl PubChem CID: 16296 IUPAC Name: pentyl benzoate SMILES: CCCCCOC(=O)C1=CC=CC=C1