accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONHOOHNHOO

CAS RN 204715-91-3

IUPAC Name: 3-[4-({[(tert-butoxy)carbonyl]amino}methyl)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-(4-{[(tert-butoxycarbonyl)amino]methyl}phenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 516.59
Molecular Formula: C30H32N2O6
InChi Key: QSPDPKLGXGMDAY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results