accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOHOHSm

CAS RN 20403-06-9

IUPAC Name: samarium(3+) trihydroxide
Synonyms: samarium(3+) trihydroxide
Molecular Weight (g/mol): 201.38
Molecular Formula: H3O3Sm
InChi Key: BCYBEIXXOVNETJ-UHFFFAOYSA-K
SMILES: [OH-].[OH-].[OH-].[Sm+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results