accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2040-04-2

CH3OOCH3CH3O

CAS RN 2040-04-2

IUPAC Name: 1-(2,6-dimethoxyphenyl)ethan-1-one
Synonyms: acetophenone, 2',6'-dimethoxy-
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: XEUGKOFTNAYMMX-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1C(C)=O
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)