accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONH(S)OOHNHOO

CAS RN 203854-47-1

IUPAC Name: 7-{[(tert-butoxy)carbonyl]amino}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)heptanoic acid
Synonyms: 7-[(tert-butoxycarbonyl)amino]-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}heptanoic acid
Molecular Weight (g/mol): 482.58
Molecular Formula: C27H34N2O6
InChi Key: SDBUQLGECIYUDT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCCC(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results