accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20362-31-6

CH3O(R)OO(R)OOOHHOHOOHOCH3OCH3

CAS RN 20362-31-6

IUPAC Name: (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(3-methoxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)methyl]oxolan-2-one
Synonyms: arctiin
Molecular Weight (g/mol): 534.56
Molecular Formula: C27H34O11
InChi Key: XOJVHLIYNSOZOO-RVDKUWPISA-N
SMILES: COC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(OC3OC(CO)C(O)C(O)C3O)C(OC)=C2)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)