CAS RN 203303-01-9
IUPAC Name:
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol
Molecular Weight (g/mol):
412.12
Molecular Formula:
C9H6F14O2
InChi Key:
XQULMKMNFZLURS-UHFFFAOYSA-N
SMILES:
OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
1
–
1
of
1
results
1
1H,1H,9H,9H-Perfluoro-1,9-nonanediol, tech. 90%
CAS: 203303-01-9 Molecular Formula: C9H6F14O2 Molecular Weight (g/mol): 412.123 MDL Number: MFCD00153275 InChI Key: XQULMKMNFZLURS-UHFFFAOYSA-N Synonym: 1h,1h,9h,9h-perfluoro-1,9-nonanediol,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol,1h,1h,9h,9h-perfluorononane-1,9-diol,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol PubChem CID: 2776243 IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononane-1,9-diol SMILES: C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O