accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OPOH3NOH3C

CAS RN 203192-98-7

IUPAC Name: diethyl (2-azaniumylethyl)phosphonate
Synonyms: diethyl 2-ammonioethylphosphonate
Molecular Weight (g/mol): 182.18
Molecular Formula: C6H17NO3P
InChi Key: JUAGHBASZWRMQH-UHFFFAOYSA-O
SMILES: CCOP(=O)(CC[NH3+])OCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results