accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20300-10-1

H2NS

CAS RN 20300-10-1

IUPAC Name: naphthalene-1-carbothioamide
Synonyms: naphthalene-1-carbothioamide
Molecular Weight (g/mol): 187.26
Molecular Formula: C11H9NS
InChi Key: DRCKUACWKCMOCB-UHFFFAOYSA-N
SMILES: NC(=S)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results