accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3NClF

CAS RN 202982-63-6

IUPAC Name: (4-chloro-2-fluorophenyl)methanaminium
Synonyms: (4-chloro-2-fluorophenyl)methanaminium
Molecular Weight (g/mol): 160.6
Molecular Formula: C7H8ClFN
InChi Key: ODMMHGMIZYLHNN-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results