accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 202865-83-6

IUPAC Name: 1-bromo-3-fluoro-5-methylbenzene
Synonyms: 1-bromo-3-fluoro-5-methylbenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: CTNFNUQREIIROB-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results