accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20120-21-2

H3COONCH3CH3

CAS RN 20120-21-2

IUPAC Name: ethyl 3-(dimethylamino)propanoate
Synonyms: ethyl 3-(dimethylamino)propanoate
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: IZMJWGJGVGRWBW-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results