accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2010-61-9

CH3OHFFFFFF

CAS RN 2010-61-9

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-ol
Synonyms: 1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-ol
Molecular Weight (g/mol): 258.16
Molecular Formula: C10H8F6O
InChi Key: AOAVZPXKNQAALI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)