CAS RN 200435-92-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2’-O,4’-C-Methyleneuridine, MedChemExpress
MedChemExpress 2’-O,4’-C-Methyleneuridine (Compound 15a) is a bicyclic nucleoside.