CAS RN 199850-67-4
CAS RN 199850-67-4
IUPAC Name:
2-{4'-bromo-[1,1'-biphenyl]-4-sulfonamido}-3-methylbutanoic acid
Synonyms:
2-{4'-bromo-[1,1'-biphenyl]-4-sulfonamido}-3-methylbutanoic acid
Molecular Weight (g/mol):
412.3
Molecular Formula:
C17H18BrNO4S
InChi Key:
GJOCABIDMCKCEG-UHFFFAOYNA-N
SMILES:
CC(C)C(NS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(Br)C=C1)C(O)=O