accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNOH

CAS RN 199590-00-6

IUPAC Name: (1-methyl-1H-indol-6-yl)methanol
Synonyms: (1-methylindol-6-yl)methanol
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: YDMPSBJXPPXTCM-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=C(CO)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results