accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 199296-61-2

H3NFFFF

CAS RN 199296-61-2

IUPAC Name: [2-fluoro-5-(trifluoromethyl)phenyl]methanaminium
Synonyms: [2-fluoro-5-(trifluoromethyl)phenyl]methanaminium
Molecular Weight (g/mol): 194.15
Molecular Formula: C8H8F4N
InChi Key: WIQWADYBGSRGCF-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC(=CC=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results